VWR®, Micro Pipettes, Disposable
Supplier: VWR
VWR®
VWRU53432-728EA 0 EUR
VWRU53432-921 VWRU53432-740 VWRU53432-783 VWRU53432-706 VWRU53432-728
VWR®, Micro Pipettes, Disposable
Pipettes Microcapillaries
Borosilicate glass.
  • Calibrated to contain
  • With permanent fill lines
  • Colour-coded
Balení: Packed in easy dispensing canisters of 250 micropipettes, plus one 38,1 cm aspirator tube. Each canister is marked with bold size designations for quick identification in the laboratory.