VWR®, Hand protectors
Supplier: VWR
VWR®
VWRU89026-680EA 0 EUR
VWRU89026-680
VWR®, Hand protectors
Hot Hand Protectors
This full-hand silicone rubber mitt is ideal for handling extremely hot or cold objects, from –57 to 260 °C, for brief periods of time. The mitt features a studded, oversized surface and internal ribs to ensure a safe grip on beakers, flasks, bottles, and similar items.
  • Ideal for handling extremely hot or cold beakers, flasks, bottles, and similar items
  • Studded, oversized surface and internal ribs ensure safe grip
  • Silicone rubber mitt is designed to comfortably fit your hand