VWR®, Sterilisation Boxes for Pipettes
Supplier: VWR
VWR®
VWRU82027-608EA 0 EUR
VWRU82027-606 VWRU82027-604 VWRU82027-608 VWRU82027-610
VWR®, Sterilisation Boxes for Pipettes
Boxes Pipette Sterilisation Containers
Pipette sterilisation boxes are designed to secure and protect pipettes during the sterilisation process.
  • Designed to secure and protect pipettes during sterilisation processes
  • Stainless steel construction ensures high corrosion resistance
  • Multiple box styles accommodate a variety of pipette types