VWR®, Aspirating Pipettes
Supplier: VWR
VWR®
VWRU414004-267EA 102 EUR
VWRU414004-266 VWRU414004-267 VWRU414004-264 VWRU414004-265
VWR®, Aspirating Pipettes
Pipettes Aspirating Pipettes
PS pipettes, sterile.
  • Can be used for a wide variety of vacuum-aspirating procedures
  • Safer alternative to glass pasteur pipettes
  • Non plugged, non graduated, non pyrogenic
  • For single use
  • Thermoformed paper/plastic packaging protects against contamination
Balení: Supplied individually wrapped in four bags of 50.