Sodium bisulphite, dried, purified, J.T.Baker®

Supplier: Avantor

3557-01EA 80.7 EUR
3557-01
Sodium bisulphite, dried, purified, J.T.Baker®
Sodium bisulphite
Formula: HNaO₃S
MW: 104,06 g/mol
Melting Pt: 300 °C
Density: 2,476 g/cm³ (20 °C)
MDL Number: MFCD00167602
CAS Number: 7631-90-5
UN: 2693
ADR: 8,III
Merck Index: 12,08731

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Identification Passes Test
Solubility in H₂O Passes Test
Thiosulfate (S₂O₃) Passes Test
Heavy Metals (as Pb) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR