Anti-ALPP Mouse Monoclonal Antibody [clone: 421CT8.1.1]

Supplier: Abgent
ABGEAM2022BEA 348 EUR
ABGEAM2022B
Anti-ALPP Mouse Monoclonal Antibody [clone: 421CT8.1.1]
Antibodies
Type: Primary
Antigen: ALPP
Clonality: Monoclonal
Clone: 421CT8.1.1
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR