Holmium(III) acetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Warning
14591.09 14591.18
14591.09EA 135 EUR
14591.09 14591.18
Holmium(III) acetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Holmium(III) acetate hydrate
Purity: 99.9% CAS No.: 312619-49-1 Molecular Formula: Ho(OOCCH3)3.xH2O Formula Weight: 342.07(anhy) Physical Form: Crystalline Aggregates MDL No.: MFCD00150648
Formula: C₆H₉HoO₆·xH₂O
MW: 342.07 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00150648
CAS Number: 312619-49-1
EINECS: 247-066-9