Iron(III) hydroxide, α-phase
Supplier: Thermo Fisher Scientific
19496.06 19496.14
19496.06EA 200 EUR
19496.06 19496.14
Iron(III) hydroxide, α-phase
Iron(III) hydroxide, alpha-phase
CAS No.: 20344-49-4 Molecular Formula: FeO(OH) Formula Weight: 88.85 Physical Form: Powder Density: 3.4-3.9 Odour: Odourless Solubility: Insoluble in water and alcohol. Soluble in mineral acids MDL No.: MFCD00064782
Applications: In purifying water, as absorbent in chemical processes, as a pigment, and as a catalyst
Formula: FeO(OH)
MW: 88.85 g/mol
Density: 3.4…3.9
Storage Temperature: Ambient
MDL Number: MFCD00064782
CAS Number: 20344-49-4
EINECS: 243-746-4
Merck Index: 14,4025