Chromium(III) phosphate tetrahydrate
Supplier: Thermo Fisher Scientific
Warning
24110.09 24110.18
24110.09EA 67.4 EUR
24110.09 24110.18
Chromium(III) phosphate tetrahydrate
Chromium(III) phosphate tetrahydrate
CAS No.: 10101-59-4 Molecular Formula: CrPO4.4H2O Formula Weight: 219.04 (146.97anhy) Physical Form: Powder Density: 2.26 MDL No.: MFCD00016049
Applications: Catalyst, pigment
Formula: CrH₈O₈P
MW: 219.026 g/mol
Density: 2.26
Storage Temperature: Ambient
MDL Number: MFCD00016049
CAS Number: 10101-59-4
EINECS: 232-141-0