Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Supplier: Thermo Scientific
|
Warning
|
35445.02
35445.04
35445.02EA
46.6
EUR
35445.02
35445.04
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
Lithium tetraphenylborate tris(1,2-dimethoxyethane) adduct
CAS No.: 75965-35-4
Molecular Formula: C24H20BLi.3C4H10O2
Formula Weight: 595.54 (326.18anhy)
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to off-white
UN No.: UN1325
MDL No.: MFCD00013311
Formula:
C₂₄H₂₀BLi.3C₄H₁₀O₂ MW: 596.531 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013311 CAS Number: 75965-35-4 ADR: 4.1,III |