Ammonium dimolybdate 4% (w/v) in aqueous solution
Supplier: Thermo Scientific
Synonyms: Ammonium molybdate (di), Diammonium dimolybdate
35578.AP 35578.K2
35578.APEA 60.7 EUR
35578.AP 35578.K2
Ammonium dimolybdate 4% (w/v) in aqueous solution
Ammonium dimolybdate
Molecular Formula: (NH4)2Mo2O7 Formula Weight: 339.95 Physical Form: Liquid MDL No.: MFCD00010882
Formula: H₈Mo₂N₂O₇
MW: 339.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010882
CAS Number: 27546-07-2