Potassium hydrogen phthalate solution_standardised 0.05 N aqueous solution volumetric solution
Supplier: Thermo Fisher Scientific
Synonyms: Potassium phthalate monobasic, Potassium biphthalate, Potassium Acid Phthalate, KHP, Phthalic Acid Monopotassium Salt, Phthalic acid potassium salt
35603.AP 35603.K2
35603.APEA 36.6 EUR
35603.AP 35603.K2
Potassium hydrogen phthalate solution_standardised 0.05 N aqueous solution volumetric solution
Potassium hydrogen phthalate
CAS No.: 877-24-7 Molecular Formula: C8H5KO4 Formula Weight: 204.23 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00013070
Formula: HOOCC₆H₄COOK
MW: 204.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM