Silica gel, granules ACS -6+18 Mesh desiccant, indicator_with
Supplier: Thermo Fisher Scientific
Danger
36737.22 36737.36
36737.22EA 60.9 EUR
36737.22 36737.36
Silica gel, granules ACS -6+18 Mesh desiccant, indicator_with
Silica gel
CAS No.: 7631-86-9 Physical Form: -6+18 Mesh Granules Sensitivity: Hygroscopic pH: 6.0 MDL No.: MFCD00011232
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4
Merck Index: 14,8490