Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced
Supplier: Thermo Fisher Scientific
38307.06 38307.14
38307.06EA 371 EUR
38307.06 38307.14
Palladium 20% on activated carbon powder wetted with ca. 50% water, eggshell, unreduced
Palladium
CAS No.: 7440-05-3 Physical Form: 900-1100m2/g Odour: Odourless MDL No.: MFCD03457879
Applications: Debenzylation
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3