Zinc 2-ethylhexanoate ∼80% (17 - 19% Zn) in mineral spirits
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Zinc(II) 2-ethylhexanoate
39561.36 39561.22 39561.0E
39561.36EA 130 EUR
39561.36 39561.22 39561.0E
Zinc 2-ethylhexanoate ∼80% (17 - 19% Zn) in mineral spirits
Zinc 2-ethylhexanoate
CAS No.: 136-53-8 Molecular Formula: C16H30O4Zn Formula Weight: 351.79 Physical Form: Liquid Density: 1.07 Appearance: Colourless UN No.: UN1268 MDL No.: MFCD00053316
Formula: C₁₆H₃₀O₄Zn
MW: 351.792 g/mol
Density: 1.07
Storage Temperature: Ambient
MDL Number: MFCD00053316
CAS Number: 136-53-8
EINECS: 205-251-1
UN: 1268
ADR: 3 III