Platinum 0.3% on alumina, pellets 2.7-3.3 mm (0.11-0.13 in), reduced
Supplier: Thermo Scientific
Synonyms: Platinium
39826.14 39826.22
39826.14EA 465 EUR
39826.14 39826.22
Platinum 0.3% on alumina, pellets 2.7-3.3 mm (0.11-0.13 in), reduced
Platinum
CAS No.: 7440-06-4 MDL No.: MFCD00011179
Applications: Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
Formula: Pt
MW: 195.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 215-691-6