Graphite, colloidal, aerosol spray lubricant

Supplier: Thermo Scientific
Danger

41775.1XEA 56 EUR
41775.1X
Graphite, colloidal, aerosol spray lubricant
Graphite, colloidal

Applications: Dry film lubricant
Air dries rapidly at room temperature and adheres well to most substrates with minimum surface preparation


Applications: Dry film lubricant

Storage Temperature: Ambient
MDL Number: MFCD00144065
UN: 1950
ADR: 2.1,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR