Poly(styrene-divinylbenzene), cross-linked_1_DVB 200-400 mesh
Supplier: Thermo Fisher Scientific
Synonyms: SDVB resin, Styrene-divinylbenzene resin, Styrene-divinylbenzene copolymer, Poly(styrene-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene
41800.14 41800.22
41800.14EA 129 EUR
41800.14 41800.22
Poly(styrene-divinylbenzene), cross-linked_1_DVB 200-400 mesh
Poly(styrene-divinylbenzene)
CAS No.: 9003-70-7 Physical Form: Beads Odour: Odourless MDL No.: MFCD00146442
Applications: For the preparation of functionalized beads used in solid-phase synthesis
Formula: [-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
MW: 234.342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00146442
CAS Number: 9003-70-7