Nickel plating solution, semi-bright finish
Supplier: Thermo Fisher Scientific
|
Danger
|
42026.AK
42026.K2
42026.AKEA
44.5
EUR
42026.AK
42026.K2
Nickel plating solution, semi-bright finish
Nickel plating solution
Physical Form: Liquid
Appearance: Green
UN No.: UN3287
MDL No.: MFCD00011137
Storage Temperature:
Ambient |
MDL Number:
MFCD00011137 UN: 3287 ADR: 6.1,III |