Poly(allylamine hydrochloride)
Supplier: Thermo Scientific
Warning
43092.09 43092.18
43092.09EA 171 EUR
43092.09 43092.18
Poly(allylamine hydrochloride)
Poly(allylamine hydrochloride)
CAS No.: 71550-12-4 Molecular Formula: [CH2CH(CH2NH2.HCl)]n Physical Form: Powder Appearance: Yellow Solubility: Soluble in water MDL No.: MFCD00084396
Applications: As a polymeric primary amine. Forms hydrogels
Formula: [CH₂CH(CH₂NH₂·HCl)]n
MW: 93.5600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084396
CAS Number: 71550-12-4
EINECS: 415-050-2