3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% (w/v) in methanol
Supplier: Thermo Scientific
Danger
Synonyms: m-TFPTAH, Trimethyl-alpha,alpha,alpha-trifluoro-m-tolylammonium Hydroxide, Trimethyl-3-trifluoromethylphenylammonium Hydroxide
43284.AC 43284.AA
43284.ACEA 578 EUR
43284.AC 43284.AA
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide 5% (w/v) in methanol
3-(Trifluoromethyl)phenyltrimethylammonium hydroxide
CAS No.: 68254-41-1 Molecular Formula: (CF3C6H4)(CH3)3NOH Formula Weight: 221.22 Physical Form: Liquid UN No.: UN2924 MDL No.: MFCD00059473
Applications: Transesterification reactions
Formula: C₁₀H₁₄F₃NO
MW: 221.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059473
CAS Number: 68254-41-1
EINECS: 200-659-6
UN: 3286
ADR: 3,II