Polyethyleneimine, M.W. 25,000, linear
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)
43896.01
43896.03
43896.01EA
750
EUR
43896.01
43896.03
Polyethyleneimine, M.W. 25,000, linear
Polyethyleneimine
CAS No.: 9002-98-6
Molecular Formula: -HN-[CH2CH2NH-]n
Physical Form: Powder
Odour: Amine-like
Melting Point: 59-60°
MDL No.: MFCD00084427
Formula:
(CH₂CH₂NH)n Melting Pt: 59…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00084427 CAS Number: 9002-98-6 |