Silver plating solution ∼28.7 g/l (metals basis)
Supplier: Thermo Fisher Scientific
Danger
44067.AK 44067.K2
44067.AKEA 53 EUR
44067.AK 44067.K2
Silver plating solution ∼28.7 g/l (metals basis)
Silver plating solution
UN No.: UN1935 MDL No.: MFCD00003397
Storage Temperature: Ambient
MDL Number: MFCD00003397
UN: 1935
ADR: 6.1,II