Ruthenium 5% on activated carbon wetted with ca. 50% water, powder, reduced
Supplier: Thermo Scientific
44338.06 44338.14 44338.22
44338.06EA 71.7 EUR
44338.06 44338.14 44338.22
Ruthenium 5% on activated carbon wetted with ca. 50% water, powder, reduced
Ruthenium
CAS No.: 7440-18-8 Odour: Odourless MDL No.: MFCD03458417
Formula: Ru
MW: 101.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1