Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness: 0.127 mm (0.005 in)
Supplier: Thermo Fisher Scientific
|
Danger
|
44952.FF
44952.FI
44952.FFEA
87.2
EUR
44952.FF
44952.FI
Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness: 0.127 mm (0.005 in)
Nitinol
Physical Form: ≈0.51g/25x25mm; Af approx. 10°C
Odour: Odourless
MDL No.: MFCD02091734