Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness: 0.38 mm (0.015 in)
Supplier: Thermo Fisher Scientific
|
Danger
|
44954.FF
44954.FI
44954.FFEA
81.8
EUR
44954.FF
44954.FI
Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness: 0.38 mm (0.015 in)
Nitinol
Physical Form: ≈1.56g/25x25mm; Af approx. 10°C
Odour: Odourless
MDL No.: MFCD02091734