Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Fisher Scientific
|
Danger
|
45492.FF
45492.FI
45492.GH
45492.FFEA
96.9
EUR
45492.FF
45492.FI
45492.GH
Nitinol, foil, superelastic, flat annealed, pickled surface, Thickness: 0.05 mm (0.002 in)
Nitinol
Physical Form: ≈0.25g/25x25mm; Af approx. 10°C
Odour: Odourless
MDL No.: MFCD02091734