Gold ≥99.999% (metals basis), wire, Premion® Ø 0.5 mm (0.02 in), annealed
Supplier: Thermo Fisher Scientific
47160.BS 47160.BW 47160.CB
47160.BSEA 140 EUR
47160.BS 47160.BW 47160.CB
Gold ≥99.999% (metals basis), wire, Premion® Ø 0.5 mm (0.02 in), annealed
Gold
Purity: 99.999% CAS No.: 7440-57-5 Physical Form: ≈3.79g/m Appearance: Gold Odour: Odourless MDL No.: MFCD00003436
Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526