Ammonia solution concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
88576.LQ 88576.LS
88576.LQEA 72.6 EUR
88576.LQ 88576.LS
Ammonia solution concentrate for dilution to 0.1 N in aqueous solution, Acculute™ standard volumetric solution
Ammonia solution
One unit makes one liter of 0.1N solution after dilution.
Physical Form: Liquid Appearance: Colourless Odour: Ammonia-like
Formula: NH₃
MW: 35.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6