Platinum 0,5% on alumina, pellets 2.7-3.3 mm (0.11-0.13 in), reduced
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
89106.14 89106.22 89106.36
89106.14EA 166 EUR
89106.14 89106.22 89106.36
Platinum 0,5% on alumina, pellets 2.7-3.3 mm (0.11-0.13 in), reduced
Platinum
CAS No.: 7440-06-4 MDL No.: MFCD00011179
Applications: Selective hydrogenations; gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 215-691-6
Merck Index: 12,07689