Platinum 5% on carbon, dry
Supplier: Thermo Fisher Scientific
Warning
A11186.06 A11186.14 A11186.03
A11186.06EA 133 EUR
A11186.06 A11186.14 A11186.03
Platinum 5% on carbon, dry
Platinum
CAS No.: 7440-06-4 UN No.: UN1325 MDL No.: MFCD00011179
Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,7529