3-(4-Hydroxy-3-methoxyphenyl)propionic acid 97%
Supplier: Thermo Scientific
Warning
Synonyms: Dihydroferulic acid, 3-(4-Hydroxy-3-methoxyphenyl)propanoic acid
A12069.03 A12069.06
A12069.03EA 69.9 EUR
A12069.03 A12069.06
3-(4-Hydroxy-3-methoxyphenyl)propionic acid 97%
3-(4-Hydroxy-3-methoxyphenyl)propionic acid
Purity: 97% CAS No.: 1135-23-5 Molecular Formula: C10H12O4 Formula Weight: 196.20 Melting Point: 87-92° Beilstein Reference: 2110370 MDL No.: MFCD00016558
Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
Melting Pt: 87…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00016558
CAS Number: 1135-23-5
EINECS: 214-489-5