2-Mercapto-4-methylpyrimidine hydrochloride 99%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Methylpyrimidine-2(1H)-thione hydrochloride
A13145.14 A13145.22
A13145.14EA 45.1 EUR
A13145.14 A13145.22
2-Mercapto-4-methylpyrimidine hydrochloride 99%
2-Mercapto-4-methylpyrimidine hydrochloride
Purity: 99% CAS No.: 6959-66-6 Molecular Formula: C5H6N2S.HCl Formula Weight: 162.64 Melting Point: ca 265° dec. Beilstein Reference: 3691520 MDL No.: MFCD00012783
Formula: C₅H₇ClN₂S
MW: 162.635 g/mol
Melting Pt: ∼ 265 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012783
CAS Number: 6959-66-6
EINECS: 230-152-5