5,7-Dihydroxy-4-methyl-2H-chromen-2-one 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 5,7-Dihydroxy-4-methylcoumarin
A14528.03 A14528.06
A14528.03EA 117 EUR
A14528.03 A14528.06
5,7-Dihydroxy-4-methyl-2H-chromen-2-one 98%
5,7-Dihydroxy-4-methyl-2H-chromen-2-one
Purity: 98% CAS No.: 2107-76-8 Molecular Formula: C10H8O4 Formula Weight: 192.17 Melting Point: 298-300° Beilstein Reference: 179133 MDL No.: MFCD00016966
Formula: C₁₀H₈O₄
MW: 192.17 g/mol
Melting Pt: 298…300 °C
Storage Temperature: Ambient
MDL Number: MFCD00016966
CAS Number: 2107-76-8
EINECS: 218-289-9