Fullerene C60 (C60/C70 70:28), powder, mixed, refined
Supplier: Thermo Scientific
Warning
Synonyms: Fullerene - C60, Buckminster-Fullerene C60, C60 Fullerene, Fullerene-C60, Bucky Ball, Footballen
41181.03 41181.06 41181.MD
41181.03EA 184 EUR
41181.03 41181.06 41181.MD
Fullerene C60 (C60/C70 70:28), powder, mixed, refined
Fullerene C60
CAS No.: 99685-96-8 Appearance: Brown/black UN No.: 1325 MDL No.: MFCD00282904
Formula: C₆₀
MW: 720.66 g/mol
Melting Pt: ? 280 °C
Density: ∼1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151408
CAS Number: 99685-96-8
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2c9ff03e3743861d242e7e023c84fcfa10a64ac&vid=53371479acfcc1403d111c470b60d1af20934c57&prid=web&clid=SAPDAM