Ammonium magnesium phosphate hexahydrate 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: AMMONIUM MAGNESIUM PHOSPHATE HEXAHYDRATE
A17497.22 A17497.36
A17497.22EA 30.6 EUR
A17497.22 A17497.36
Ammonium magnesium phosphate hexahydrate 98%
Ammonium magnesium phosphate hexahydrate
Purity: 98% CAS No.: 13478-16-5 Molecular Formula: NH4MgPO4.6H2O Formula Weight: 245.41 (137.32anhy) MDL No.: MFCD00150461
Formula: NH₄MgPO₄·6H₂O
MW: 245.41 g/mol (137.32 anhy)
Density: 1.711
Storage Temperature: Ambient
MDL Number: MFCD00150461
CAS Number: 13478-16-5
EINECS: 232-075-2