1,4-Benzoquinone dioxime 95%
Supplier: Thermo Scientific
Warning
Synonyms: p-Quinone Dioxime
A17532.0B A17532.18 A17532.30
A17532.0BEA 445 EUR
A17532.0B A17532.18 A17532.30
1,4-Benzoquinone dioxime 95%
1,4-Benzoquinone dioxime
Purity: 95% CAS No.: 105-11-3 Molecular Formula: C6H6N2O2 Formula Weight: 138.13 Melting Point: ca 223° dec. Beilstein Reference: 2043234 MDL No.: MFCD00063636
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: ∼ 223 °C (dec.)
Density: 1.490
Storage Temperature: Ambient
MDL Number: MFCD00063636
CAS Number: 105-11-3
EINECS: 203-271-5