1-Allyl-3-(2-hydroxyethyl)-2-thiourea 97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1-Allyl-3-(2-hydroxyethyl)thiourea
A18161.18
A18161.30
A18161.18EA
46.7
EUR
A18161.18
A18161.30
1-Allyl-3-(2-hydroxyethyl)-2-thiourea 97%
1-Allyl-3-(2-hydroxyethyl)-2-thiourea
Purity: 97%
CAS No.: 105-81-7
Molecular Formula: C6H12N2OS
Formula Weight: 160.24
Melting Point: 77-79°
MDL No.: MFCD00002838
Formula:
C₆H₁₂N₂OS Melting Pt: 77…79 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002838 CAS Number: 105-81-7 EINECS: 203-334-7 |