Acid green 1
Supplier: Thermo Fisher Scientific
Synonyms: Naphthol Green B, Iron, tris(5,6-dihydro-5,6-dioxo-2-naphthalenesulphonic acid 5-oximato)-,trisodium salt
A18268.14 A18268.22
A18268.14EA 25.5 EUR
A18268.14 A18268.22
Acid green 1
Acid green 1
CAS No.: 19381-50-1 Molecular Formula: C30H15FeN3Na3O15S3 Formula Weight: 878.47 MDL No.: MFCD00003886
Formula: C₃₀H₁₈FeN₃O₁₅S₃
MW: 878.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003886
CAS Number: 19381-50-1
EINECS: 243-010-2