Glutaconaldehydedianil hydrochloride mixed isomers 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: N-(5-Anilino-2,4-pentadienylidene)aniline hydrochloride, N-[5-(Phenylamino)-2,4-pentadienylidene]aniline monohydrochloride, N-((2E,4E)-5-(Phenylamino)penta-2,4-dien-1-ylidene)aniline hydrochloride
A18390.06 A18390.14 A18390.22
A18390.06EA 83.3 EUR
A18390.06 A18390.14 A18390.22
Glutaconaldehydedianil hydrochloride mixed isomers 98%
Glutaconaldehydedianil hydrochloride
Purity: 98% CAS No.: 1497-49-0 Molecular Formula: C6H5NHCH=CHCH=CHCH=NC6H5.HCl Formula Weight: 284.79 Melting Point: ca 124-132° Beilstein Reference: 3916924 MDL No.: MFCD00012630
Formula: C₁₇H₁₆N₂
MW: 284.79 g/mol
Melting Pt: ∼ 124…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00012630
CAS Number: 1497-49-0
EINECS: 216-094-3