2-Nitrobenzylamine hydrochloride 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: (2-Nitrophenyl)methanamine hydrochloride
A18900.06 A18900.03
A18900.06EA 164 EUR
A18900.06 A18900.03
2-Nitrobenzylamine hydrochloride 98%
2-Nitrobenzylamine hydrochloride
Purity: 98% CAS No.: 24835-08-3 Molecular Formula: C7H8N2O2.HCl Formula Weight: 188.62 Melting Point: 250° dec. MDL No.: MFCD00136280
Formula: C₇H₈N₂O₂·HCl
MW: 188.62 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00136280
CAS Number: 24835-08-3