Ethyl-3-hexenoate 98%
Supplier: Thermo Scientific
Warning
Synonyms: Ethyl hex-3-enoate
A19734.06 A19734.14
A19734.06EA 28.3 EUR
A19734.06 A19734.14
Ethyl-3-hexenoate 98%
Ethyl-3-hexenoate
Purity: 98% CAS No.: 2396-83-0 Molecular Formula: CH3CH2CH=CHCH2CO2CH2CH3 Formula Weight: 142.20 Flash Point: 59°(139°F) Refractive Index: 1.4260 Boiling Point: 63-64°/12mm UN No.: UN3272 MDL No.: MFCD00036524
Formula: CH₃CH₂CH=CHCH₂CO₂CH₂CH₃
MW: 142.20 g/mol
Boiling Pt: 63…64 °C (12 mmHg)
Density: 0.89
Flash Pt: 59 °C (139 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036524
CAS Number: 2396-83-0
EINECS: 219-257-7
ADR: 3,III