4,5-Dibromothiophene-2-carboxaldehyde 98%
Supplier: Thermo Scientific
Warning
Synonyms: 4,5-Dibromothiophene-2-carbaldehyde
A19788.06 A19788.14
A19788.06EA 88.7 EUR
A19788.06 A19788.14
4,5-Dibromothiophene-2-carboxaldehyde 98%
4,5-Dibromothiophene-2-carboxaldehyde
Purity: 98% CAS No.: 38071-22-6 Molecular Formula: C5H2Br2OS Formula Weight: 269.94 Melting Point: 79-81° MDL No.: MFCD00225242
Formula: C₅H₂Br₂OS
MW: 269.94 g/mol
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00225242
CAS Number: 38071-22-6