4-Hydrazinobenzenesulphonamide hydrochloride 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 4-Hydrazinylbenzenesulphonamide hydrochloride, (4-sulphamoylphenyl)hydrazine HCl, 4-Sulphonamidophenylhydrazine Hydrochloride
B20015.09 B20015.18 B20015.88
B20015.09EA 45.5 EUR
B20015.09 B20015.18 B20015.88
4-Hydrazinobenzenesulphonamide hydrochloride 97%
4-Hydrazinobenzenesulphonamide hydrochloride
Purity: 97% CAS No.: 17852-52-7 Molecular Formula: C9H9N3O2S.HCl Formula Weight: 223.68 Melting Point: 217-219° dec. MDL No.: MFCD00052262
Formula: C₆H₉N₃O₂S
MW: 223.68 g/mol
Melting Pt: 217…219 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052262
CAS Number: 17852-52-7