Methyl-3-(methylamino)but-2-enoate 97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Methyl 3-(methylamino)but-2-enoate
B20199.06 B20199.14
B20199.06EA 83.1 EUR
B20199.06 B20199.14
Methyl-3-(methylamino)but-2-enoate 97%
Methyl-3-(methylamino)but-2-enoate
Purity: 97% CAS No.: 13412-12-9 Molecular Formula: CH3C(NHCH3)=CHCO2CH3 Formula Weight: 129.16 Melting Point: 60-63° MDL No.: MFCD00027383
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 60…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00027383
CAS Number: 13412-12-9