Ethylene glycol ethyl methyl ether 97% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.01%
B20819.09
B20819.18
B20819.09EA
43.4
EUR
B20819.09
B20819.18
Ethylene glycol ethyl methyl ether 97% stabilised
Ethylene glycol ethyl methyl ether
Stab. with 0.01% BHT
Purity: 97%
CAS No.: 5137-45-1
Molecular Formula: C5H12O2
Formula Weight: 104.15
Flash Point: 2°(36°F)
Refractive Index: 1.3870
Boiling Point: 100-102°
UN No.: UN3271
MDL No.: MFCD00048580
Formula:
C₅H₁₂O₂ Boiling Pt: 100…102 °C Density: 0.850 Flash Pt: 2 °C (36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00048580 CAS Number: 5137-45-1 EINECS: 225-893-6 UN: 3271 ADR: 3,II |