D-(-)-Camphorsulphonylimine ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(1S)-(-)-Camphorsulphonylimine, (-)-10-Camphorsulphonimine, (3aS)-(-)-4,5,6,7-Tetrahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole 2,2-Dioxide
B20823.03
B20823.06
B20823.14
B20823.03EA
31.9
EUR
B20823.03
B20823.06
B20823.14
D-(-)-Camphorsulphonylimine ≥98%
D-(-)-Camphorsulphonylimine
Purity: 98+%
CAS No.: 60886-80-8
Molecular Formula: C10H15NO2S
Formula Weight: 213.30
Melting Point: 226-228°
Beilstein Reference: 85296
MDL No.: MFCD00064576
Formula:
C₁₀H₁₅NO₂S Melting Pt: 226…228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064576 CAS Number: 60886-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...