2,6-Diaminopimelic acid 96%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,5-Diaminoheptanedioic Acid, 2,6-Diaminoheptanedioic acid

B22391.03 B22391.06
B22391.03EA 54.9 EUR
B22391.03 B22391.06
2,6-Diaminopimelic acid 96%
2,6-Diaminopimelic acid

Purity: 96% CAS No.: 583-93-7 Molecular Formula: C7H14N2O4 Formula Weight: 190.2 Physical Form: Powder Appearance: White Melting Point: 300° MDL No.: MFCD00002637

Formula: C₇H₁₄N₂O₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00002637
CAS Number: 583-93-7
EINECS: 209-524-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR