4-Cyanostyrene 97% stabilised

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 4-Ethenylbenzonitrile

Stabilization: 0.05% Stabilised with 4-tert-butylpyrocatechol (TBC)

B24395.03 B24395.06 B24395.14
B24395.03EA 80.1 EUR
B24395.03 B24395.06 B24395.14
4-Cyanostyrene 97% stabilised
4-Cyanostyrene

Stab. with 0.05% 4-tert-butyl catechol


Purity: 97% CAS No.: 3435-51-6 Molecular Formula: C9H7N Formula Weight: 129.16 Storage Temperature: 2°C to 8°C Refractive Index: 1.5775 Boiling Point: 92-93°/3mm UN No.: UN3276 MDL No.: MFCD00080445

Formula: C₉H₇N
MW: 129.162 g/mol
Boiling Pt: 92…93 °C (3 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00080445
CAS Number: 3435-51-6
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR