cis-5-Norbornene endo-2,3-dicarboxylic anhydride 97%
Supplier: Thermo Scientific
Danger
Synonyms: (3aR,4S,7R,7aS)-rel-3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione, Carbic anhydride
B24951.06 B24951.14 B24951.22
B24951.06EA 35.9 EUR
B24951.06 B24951.14 B24951.22
cis-5-Norbornene endo-2,3-dicarboxylic anhydride 97%
cis-5-Norbornene endo-2,3-dicarboxylic anhydride
Purity: 97% CAS No.: 129-64-6 Molecular Formula: C9H8O3 Formula Weight: 164.16 Melting Point: 164-167° Beilstein Reference: 83657 MDL No.: MFCD00151106
Formula: C₉H₈O₃
MW: 164.16 g/mol
Melting Pt: 164…167 °C
Density: 1.417
Storage Temperature: Ambient
MDL Number: MFCD00151106
CAS Number: 129-64-6
EINECS: 204-957-7
Merck Index: 14,1796